Assay Design



Gene Transcripts

Gene Symbol
Trim75
Entrez Gene ID
333307
Gene Name
tripartite motif-containing 75
Gene Aliases
Gm794
Chromosome Location
Chr.8: 64980588 - 64987669 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033429.2 NP_001028601.1 1 - 2 44 87
GenBank mRNA AK136018.1 - 1 - 2 46 87

View Full Product Details