Assay Design



Gene Transcripts

Gene Symbol
Nlrp6
Entrez Gene ID
101613
Gene Name
NLR family, pyrin domain containing 6
Gene Aliases
AI504961, Avr, Nalp6, Navr, Navr/Avr, Pypaf5
Chromosome Location
Chr.7: 140920902 - 140929192 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133946.2 NP_598707.2 1 - 2 183 103
GenBank mRNA BB869957.1 - 1 - 2 183 102
FJ222825.1 - 1 - 2 132 103

View Full Product Details