Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085540.2 | NP_001079009.1 | 1 - 2 | 280 | 106 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085541.1 | NP_001079010.1 | 1 - 2 | 280 | 106 |