Assay Design
Gene Transcripts
- Gene Symbol
- Vmn2r6
- Entrez Gene ID
-
667069
- Gene Name
- vomeronasal 2, receptor 6
- Gene Aliases
-
EG620718, EG667069
- Chromosome Location
-
Chr.3: 64537501 - 64565434 on Build GRCm38
- Gene Symbol
- Vmn2r7
- Entrez Gene ID
-
319217
- Gene Name
- vomeronasal 2, receptor 7
- Gene Aliases
-
4933425M15Rik
- Chromosome Location
-
Chr.3: 64690660 - 64719602 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175674.3 |
NP_783605.2 |
4 - 5 |
|
87 |
View Full Product Details