Assay Design



Gene Transcripts

Gene Symbol
Vmn2r31
Entrez Gene ID
100042591
Gene Name
vomeronasal 2, receptor 31
Gene Aliases
-
Chromosome Location
Chr.7: 7383985 - 7399627 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105062.1 NP_001098532.1 1 - 2 199 101

Gene Symbol
Vmn2r33
Entrez Gene ID
624512
Gene Name
vomeronasal 2, receptor 33
Gene Aliases
-
Chromosome Location
Chr.7: 7550967 - 7566786 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105065.2 NP_001098535.2 1 - 2 199 101

Gene Symbol
Vmn2r38
Entrez Gene ID
434110
Gene Name
vomeronasal 2, receptor 38
Gene Aliases
-
Chromosome Location
Chr.7: 9074796 - 9097765 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105070.1 NP_001098540.1 1 - 2 199 101

Gene Symbol
Vmn2r44
Entrez Gene ID
434113
Gene Name
vomeronasal 2, receptor 44
Gene Aliases
EG434113
Chromosome Location
Chr.7: 8367460 - 8383238 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105074.1 NP_001098544.1 1 - 2 199 101

View Full Product Details