Assay Design



Gene Transcripts

Gene Symbol
B3gnt5
Entrez Gene ID
108105
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Aliases
beta3GnT5
Chromosome Location
Chr.16: 19760192 - 19772758 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159407.1 NP_001152879.1 4 - 5 742 103
NM_001159408.1 NP_001152880.1 3 - 4 539 103
GenBank mRNA AK037166.1 - 3 - 4 566 103

View Full Product Details