Assay Design



Gene Transcripts

Gene Symbol
Serpinb6a
Entrez Gene ID
20719
Gene Name
serine (or cysteine) peptidase inhibitor, clade B, member 6a
Gene Aliases
4930482L21Rik, D330015H01Rik, PI-6, Serpinb6, Spi3, ovalbumin
Chromosome Location
Chr.13: 33917918 - 34002794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164117.1 NP_001157589.1 2 - 3 333 77
GenBank mRNA AK154700.1 - 2 - 3 229 77
BC006766.1 - 2 - 3 246 77

View Full Product Details