Assay Design
Gene Transcripts
- Gene Symbol
- Gm5803
- Entrez Gene ID
-
545091
- Gene Name
- predicted gene 5803
- Gene Aliases
-
EG545091
- Chromosome Location
-
Chr.15: 22713820 - 22714984 on Build GRCm38
- Gene Symbol
- Gm10052
- Entrez Gene ID
-
654467
- Gene Name
- heterogeneous nuclear ribonucleoprotein A1 pseudogene
- Gene Aliases
-
ENSMUSG00000058922, XM_001481145
- Chromosome Location
-
Chr.9: 123688848 - 123690275 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002885.3 |
|
1 - 1 |
282 |
96 |
- Gene Symbol
- Gm5643
- Entrez Gene ID
-
434858
- Gene Name
- heterogeneous nuclear ribonucleoprotein A1 pseudogene
- Gene Aliases
-
EG434858
- Chromosome Location
-
Chr.X: 142297505 - 142299237 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002883.1 |
|
1 - 1 |
262 |
96 |
- Gene Symbol
- Hnrnpa1
- Entrez Gene ID
-
15382
- Gene Name
- heterogeneous nuclear ribonucleoprotein A1
- Gene Aliases
-
HDP-1, Hdp, Hnrpa1, hnRNP A1, hnrnp-A1
- Chromosome Location
-
Chr.15: 103240397 - 103246698 on Build GRCm38
View Full Product Details