Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010003.2 | NP_034133.2 | 2 - 3 | 382 | 91 |
GenBank mRNA | AF047726.1 | - | 2 - 3 | 333 | 91 |
BC057911.1 | - | 2 - 3 | 343 | 91 | |
BC139290.1 | - | 2 - 3 | 382 | 91 | |
BC139291.1 | - | 2 - 3 | 382 | 91 |