Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025338.4 | NP_079614.1 | 3 - 4 | 998 | 74 |
GenBank mRNA | AK002737.1 | - | 3 - 4 | 617 | 74 |
AK003442.1 | - | 3 - 4 | 622 | 74 | |
AK033452.1 | - | 3 - 4 | 605 | 74 | |
AK144719.1 | - | 3 - 4 | 590 | 74 | |
AK158165.1 | - | - | 607 | 74 | |
AK166049.1 | - | 3 - 4 | 602 | 74 | |
BC011245.1 | - | - | 609 | 74 |