Assay Design



Gene Transcripts

Gene Symbol
Krtap3-3
Entrez Gene ID
66380
Gene Name
keratin associated protein 3-3
Gene Aliases
2310020D06Rik, KAP3.3, KRTAP3.3
Chromosome Location
Chr.11: 99550132 - 99550863 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025524.2 NP_079800.1 1 - 1 548 103
GenBank mRNA AK009052.1 - 1 - 1 552 103
AK009410.1 - 1 - 1 550 103
AK010142.1 - 1 - 1 549 103

View Full Product Details