Assay Design
Gene Transcripts
- Gene Symbol
- Commd5
- Entrez Gene ID
-
66398
- Gene Name
- COMM domain containing 5
- Gene Aliases
-
2310065H03Rik, AI854466, D15Ertd81e, Hcarg
- Chromosome Location
-
Chr.15: 76899941 - 76901297 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_025536.2 |
NP_079812.1 |
1 - 2 |
49 |
93 |
GenBank mRNA |
AK010045.1 |
- |
1 - 2 |
50 |
93 |
View Full Product Details