Assay Design
Gene Transcripts
- Gene Symbol
- Sectm1b
- Entrez Gene ID
-
58210
- Gene Name
- secreted and transmembrane 1B
- Gene Aliases
-
1810003C24Rik, K12, Sectm1
- Chromosome Location
-
Chr.11: 121053423 - 121063569 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026907.3 |
NP_081183.3 |
1 - 2 |
54 |
112 |
GenBank mRNA |
AK029082.1 |
- |
1 - 2 |
55 |
112 |
View Full Product Details