Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271411.1 | NP_001258340.1 | 4 - 5 | 820 | 110 |
NM_001271412.1 | NP_001258341.1 | 4 - 5 | 750 | 110 | |
NM_001271413.1 | NP_001258342.1 | 4 - 5 | 659 | 110 | |
NM_001271414.1 | NP_001258343.1 | 4 - 5 | 747 | 110 | |
NM_028728.3 | NP_083004.1 | 4 - 5 | 677 | 110 | |
GenBank mRNA | AF361364.1 | - | 4 - 5 | 754 | 110 |
AK014802.2 | - | 4 - 5 | 678 | 110 | |
AK048992.1 | - | 4 - 5 | 661 | 110 | |
AK150635.1 | - | 4 - 5 | 729 | 110 | |
AK151187.1 | - | 4 - 5 | 729 | 110 | |
AK151884.1 | - | 4 - 5 | 729 | 110 | |
AK152262.1 | - | 4 - 5 | 729 | 110 | |
AK152499.1 | - | 4 - 5 | 729 | 110 | |
AK152688.1 | - | 4 - 5 | 729 | 110 | |
AK155106.1 | - | 4 - 5 | 674 | 110 | |
AK162939.1 | - | 4 - 5 | 731 | 110 | |
AK169918.1 | - | 4 - 5 | 674 | 110 | |
AK170098.1 | - | 4 - 5 | 764 | 110 | |
AK170355.1 | - | 4 - 5 | 727 | 110 | |
AK170442.1 | - | 4 - 5 | 729 | 110 | |
AK170545.1 | - | 4 - 5 | 615 | 110 | |
AK170593.1 | - | 4 - 5 | 685 | 110 | |
AK170677.1 | - | 4 - 5 | 685 | 110 | |
AK170811.1 | - | 4 - 5 | 685 | 110 | |
AK170883.1 | - | 4 - 5 | 728 | 110 | |
AK171082.1 | - | 4 - 5 | 790 | 110 | |
AK171086.1 | - | 4 - 5 | 698 | 110 | |
AK171201.1 | - | 4 - 5 | 659 | 110 |