Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029017.2 | NP_083293.1 | 4 - 5 | 430 | 87 |
GenBank mRNA | BC029173.1 | - | - | 410 | 87 |
BC038065.1 | - | 4 - 5 | 401 | 87 | |
BC087953.1 | - | 4 - 5 | 424 | 87 | |
CX122942.1 | - | 4 - 5 | 409 | 87 |