Assay Design



Gene Transcripts

Gene Symbol
Lrrc48
Entrez Gene ID
74665
Gene Name
leucine rich repeat containing 48
Gene Aliases
4930449E07Rik
Chromosome Location
Chr.11: 60353206 - 60394342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029044.2 NP_083320.1 13 - 14 1641 76
GenBank mRNA AK015430.1 - 13 - 14 1643 76
BC018412.1 - 13 - 14 1654 76

View Full Product Details