Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029092.3 | NP_083368.1 | 1 - 2 | 279 | 93 |
GenBank mRNA | AK005047.1 | - | 1 - 2 | 283 | 93 |
AK087991.1 | - | 1 - 2 | 272 | 93 | |
AK135426.1 | - | 1 - 2 | 285 | 93 | |
AK148219.1 | - | - | 250 | 93 | |
AK166781.1 | - | 1 - 2 | 292 | 93 | |
BC023147.1 | - | 1 - 2 | 259 | 93 | |
BC106131.1 | - | 1 - 2 | 261 | 93 |