Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029478.3 | NP_083754.2 | 6 - 7 | 689 | 96 |
GenBank mRNA | AB047550.1 | - | 6 - 7 | 701 | 96 |
AB076609.1 | - | 5 - 6 | 580 | 96 | |
AK016312.1 | - | 6 - 7 | 706 | 96 | |
AK043091.1 | - | 6 - 7 | 690 | 96 | |
AK049559.1 | - | 6 - 7 | 682 | 96 | |
AK077443.1 | - | 6 - 7 | 689 | 96 | |
AK159468.1 | - | 6 - 7 | 704 | 96 | |
AK167173.1 | - | 6 - 7 | 697 | 96 | |
AK167202.1 | - | 6 - 7 | 697 | 96 | |
AK189413.1 | - | 2 - 3 | 269 | 96 | |
AK189549.1 | - | 2 - 3 | 208 | 96 | |
AK216058.1 | - | 2 - 3 | 268 | 96 | |
AX775963.1 | - | 6 - 7 | 659 | 96 | |
BC004013.1 | - | 6 - 7 | 683 | 96 |