Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053079.2 | NP_444309.2 | 9 - 10 | 751 | 106 |
GenBank mRNA | AF205540.1 | - | 9 - 10 | 750 | 106 |
AK085471.1 | - | 7 - 8 | 1858 | 106 | |
AK136428.1 | - | 9 - 10 | 752 | 106 | |
BC116248.1 | - | 9 - 10 | 741 | 106 | |
BC116249.1 | - | 9 - 10 | 741 | 106 |