Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130450.2 | NP_569717.1 | 1 - 2 | 109 | 68 |
GenBank mRNA | AB072039.1 | - | 1 - 2 | 56 | 68 |
AK029029.1 | - | 1 - 2 | 111 | 68 | |
AK087440.1 | - | 1 - 2 | 129 | 68 | |
AK167373.1 | - | 1 - 2 | 75 | 68 | |
AY053453.1 | - | 1 - 2 | 90 | 68 | |
BC040097.1 | - | 1 - 2 | 51 | 68 | |
BC051041.1 | - | 1 - 2 | 106 | 68 | |
BC100576.1 | - | 1 - 2 | 51 | 68 |