Assay Design



Gene Transcripts

Gene Symbol
Igflr1
Entrez Gene ID
101883
Gene Name
IGF-like family receptor 1
Gene Aliases
F630003I17Rik, Tmem149
Chromosome Location
Chr.7: 30565427 - 30567962 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145580.2 NP_663555.2 4 - 5 807 80
GenBank mRNA AK154129.1 - 4 - 5 808 80
AW553050.2 - 1 - 2 80
BC019733.1 - 3 - 4 848 80

View Full Product Details