Assay Design



Gene Transcripts

Gene Symbol
Prss30
Entrez Gene ID
30943
Gene Name
protease, serine 30
Gene Aliases
2010015P21Rik, Disp, Disp1, Tmprss8
Chromosome Location
Chr.17: 23972127 - 23980572 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013921.3 NP_038949.2 1 - 2 103 86
GenBank mRNA AJ243866.1 - 1 - 2 66 86
BC010970.1 - 1 - 2 96 86
BC040348.1 - 1 - 2 103 86

View Full Product Details