Assay Design



Gene Transcripts

Gene Symbol
Ms4a13
Entrez Gene ID
73466
Gene Name
membrane-spanning 4-domains, subfamily A, member 13
Gene Aliases
1700060E18Rik, Tetm4.2
Chromosome Location
Chr.19: 11169418 - 11196755 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198224.3 NP_937867.2 1 - 2 174 101
GenBank mRNA AF544402.1 - 1 - 2 179 101
BC089483.1 - 1 - 2 174 101

View Full Product Details