Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201227.3 | NP_957679.1 | 1 - 2 | 455 | 108 |
GenBank mRNA | AB070694.1 | - | 1 - 2 | 312 | 108 |
AY387409.1 | - | 1 - 2 | 451 | 108 | |
BC115659.1 | - | 1 - 2 | 404 | 108 | |
BC115660.2 | - | 1 - 2 | 108 |