Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011590.2 | NP_035720.1 | 6 - 6 | 852 | 104 |
GenBank mRNA | AK003129.1 | - | 6 - 6 | 853 | 104 |
BC010830.1 | - | 6 - 6 | 840 | 104 | |
BC098216.1 | - | 6 - 6 | 849 | 104 |