Assay Design



Gene Transcripts

Gene Symbol
Pcdhga3
Entrez Gene ID
93711
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
-
Chromosome Location
Chr.18: 37674335 - 37841873 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033586.2 NP_291064.1 1 - 1 1452 100
GenBank mRNA AY013797.1 - 1 - 1 1291 100
BC060691.1 - 1 - 1 1452 100

View Full Product Details