Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017372.3 | NP_059068.1 | 1 - 1 | 153 | 102 |
GenBank mRNA | AK148516.1 | - | 1 - 1 | 154 | 102 |
AK150998.1 | - | 1 - 1 | 156 | 102 | |
AK153244.1 | - | 1 - 1 | 156 | 102 | |
AK153475.1 | - | 1 - 1 | 154 | 102 | |
AK159276.1 | - | 1 - 1 | 203 | 102 | |
AK159640.1 | - | 1 - 1 | 150 | 102 | |
BC002069.1 | - | 1 - 1 | 145 | 102 | |
BC054463.1 | - | 1 - 1 | 145 | 102 |