Assay Design



Gene Transcripts

Gene Symbol
Kcnk15
Entrez Gene ID
241769
Gene Name
potassium channel, subfamily K, member 15
Gene Aliases
KCNK11, KCNK14, KT3.3, TASK5
Chromosome Location
Chr.2: 163853750 - 163858874 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030292.1 NP_001025463.1 1 - 1 153 102
GenBank mRNA BC147255.1 - 1 - 1 252 102
BC147256.1 - 1 - 1 252 102

View Full Product Details