Assay Design
Gene Transcripts
- Gene Symbol
- Defa-rs12
- Entrez Gene ID
-
13221
- Gene Name
- defensin, alpha, related sequence 12
- Gene Aliases
-
CRS4C-5, Defcr-rs12
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_007846.1 |
NP_031872.1 |
- |
149 |
92 |
View Full Product Details