Assay Design



Gene Transcripts

Gene Symbol
ND3
Entrez Gene ID
17718
Gene Name
NADH dehydrogenase subunit 3
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK139029.1 - 1 - 1 46 134
NC_005089_ND3.0 - - 46 134

View Full Product Details