Assay Design
Gene Transcripts
- Gene Symbol
- Mir34c
- Entrez Gene ID
-
723932
- Gene Name
- microRNA 34c
- Gene Aliases
-
Mirn34c, mir-34c, mmu-mir-34c
- Chromosome Location
-
Chr.9: 51103034 - 51103110 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029654.1 |
|
1 - 1 |
38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
B cell translocation gene 4 |
Btg4 |
C86116, PC3B |
|
Anti-sense |
Chr. - 51102265 - 51119700 on Build GRCm38 |
Downstream |
microRNA 34b |
Mir34b |
Mirn34b, mir-34b, mmu-mir-34b |
452.0
|
Sense |
Chr. - 51103562 - 51103645 on Build GRCm38 |
View Full Product Details