Assay Design



Gene Transcripts

Gene Symbol
Mblac1
Entrez Gene ID
330216
Gene Name
metallo-beta-lactamase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 138194314 - 138195621 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177878.3 NP_808546.1 1 - 1 657 76
GenBank mRNA AK049381.1 - 1 - 1 658 76
BC038925.1 - 1 - 1 638 76

View Full Product Details