Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243064.1 | NP_001229993.1 | 2 - 2 | 1981 | 84 |
NM_007616.4 | NP_031642.1 | 3 - 3 | 1885 | 84 | |
GenBank mRNA | AB029929.1 | - | 3 - 3 | 1848 | 84 |
AB029930.1 | - | 2 - 2 | 1896 | 84 | |
AK003489.1 | - | 3 - 3 | 1831 | 84 | |
AK010839.1 | - | 1 - 1 | 201 | 84 | |
AK028738.1 | - | 3 - 3 | 1819 | 84 | |
AK035592.1 | - | 3 - 3 | 1823 | 84 | |
AK090074.1 | - | 2 - 2 | 1901 | 84 | |
BC038280.1 | - | 3 - 3 | 1792 | 84 | |
BC052859.1 | - | 3 - 3 | 1795 | 84 |