Assay Design
Gene Transcripts
- Gene Symbol
- Defa-ps12
- Entrez Gene ID
-
654452
- Gene Name
- defensin, alpha, pseudogene 12
- Gene Aliases
-
Defb46-ps, EG654452
- Chromosome Location
-
Chr.8: 19210462 - 19212760 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002878.2 |
|
2 - 2 |
126 |
78 |
GenBank mRNA |
DQ012047.1 |
- |
2 - 2 |
126 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
defensin beta 4 |
Defb4 |
2310001F05Rik, BD-4 |
-11.1kb
|
Sense |
Chr. - 19198704 - 19201547 on Build GRCm38 |
Downstream |
defensin beta 6 |
Defb6 |
BD-6 |
12.7kb
|
Sense |
Chr. - 19225478 - 19228209 on Build GRCm38 |
View Full Product Details