Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010293.3 | NP_034423.2 | 1 - 1 | 1421 | 93 |
GenBank mRNA | AF117733.1 | - | 1 - 1 | 1421 | 93 |
AK076631.1 | - | 1 - 1 | 1397 | 93 | |
BC050968.1 | - | 1 - 1 | 1410 | 93 | |
BC119574.2 | - | 1 - 1 | 1402 | 93 |