Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_045542.1 | 2 - 3 | 458 | 111 | |
GenBank mRNA | AK044178.1 | - | 2 - 3 | 393 | 111 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | LKAAEAR motif containing 1 (IKAAEAR murine motif) | Lkaaear1 | 4930526D03Rik, Gm692 | -17.2kb | Anti-sense | Chr. - 181696793 - 181698468 on Build GRCm38 |
Downstream | myelin transcription factor 1 | Myt1 | Nzf2, Nzf2a, Nzf2b, Nztf2 | 46.8kb | Sense | Chr. - 181763332 - 181827781 on Build GRCm38 |