Assay Design



Gene Transcripts

Gene Symbol
Crygc
Entrez Gene ID
12966
Gene Name
crystallin, gamma C
Gene Aliases
Cryg-5
Chromosome Location
Chr.1: 65071525 - 65073532 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082573.2 NP_001076042.1 2 - 3 283 102
GenBank mRNA EF426302.1 - 2 - 3 272 102

View Full Product Details