Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_073425.1 | 4 - 5 | 1366 | 75 | |
GenBank mRNA | AK134925.1 | - | 4 - 5 | 624 | 75 |
AK158925.1 | - | 4 - 5 | 1366 | 75 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | chloride intracellular channel 5 | Clic5 | 5730531E12Rik, B330005L24, Gm322, jbg | -358.3kb | Anti-sense | Chr. - 44134678 - 44280168 on Build GRCm38 |
Downstream | runt related transcription factor 2, opposite strand 2 | Runx2os2 | 9530072K05Rik | 61.6kb | Anti-sense | Chr. - 44701390 - 44714098 on Build GRCm38 |