Assay Design
Gene Transcripts
- Gene Symbol
- Osgepl1
- Entrez Gene ID
-
72085
- Gene Name
- O-sialoglycoprotein endopeptidase-like 1
- Gene Aliases
-
2610001M19Rik, AA416452
- Chromosome Location
-
Chr.1: 53312897 - 53326343 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104360.1 |
|
5 - 6 |
1352 |
70 |
GenBank mRNA |
BC030671.1 |
- |
5 - 6 |
1322 |
70 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY464147.1 |
- |
1 - 2 |
155 |
|
Chr. 1 - 53321080 - 53326174 [+] on Build GRCm38 |
70 |
GenBankmRNA |
AI504862.1 |
- |
2 - 3 |
|
|
Chr. 1 - 53320239 - 53326152 [-] on Build GRCm38 |
70 |
GenBankmRNA |
BB043703.2 |
- |
2 - 3 |
436 |
|
Chr. 1 - 53319997 - 53326150 [+] on Build GRCm38 |
70 |
GenBankmRNA |
BG144363.1 |
- |
2 - 3 |
|
|
Chr. 1 - 53320131 - 53326160 [-] on Build GRCm38 |
70 |
GenBankmRNA |
BB809868.1 |
- |
2 - 3 |
157 |
|
Chr. 1 - 53320276 - 53326149 [+] on Build GRCm38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ORM1-like 1 (S. cerevisiae) |
Ormdl1 |
C730042F17Rik |
-10.8kb
|
Sense |
Chr. - 53297095 - 53310245 on Build GRCm38 |
Downstream |
asparagine synthetase domain containing 1 |
Asnsd1 |
2210409M21Rik |
21.3kb
|
Anti-sense |
Chr. - 53344617 - 53352752 on Build GRCm38 |
View Full Product Details