Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104433.1 | 3 - 4 | 495 | 86 | |
GenBank mRNA | AF401531.2 | - | 3 - 4 | 400 | 86 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | thymidine kinase 2, mitochondrial | Tk2 | AU024611, mt-TK | -12.9kb | Anti-sense | Chr. - 104226691 - 104248574 on Build GRCm38 |
Downstream | CKLF-like MARVEL transmembrane domain containing 2A | Cmtm2a | 1700001K04Rik, 1700041N15Rik, 1700063K20Rik, AI449770, ARR19, C32, CKLF3, CKLF4, CKLF5, Cklf, Cklf1, Cklfsf2-1b, Cklfsf2a, UCK-1 | 18.8kb | Anti-sense | Chr. - 104281042 - 104293181 on Build GRCm38 |