Assay Design



Gene Transcripts

Gene Symbol
LIM2
Entrez Gene ID
101110785
Gene Name
lens intrinsic membrane protein 2
Gene Aliases
-
Chromosome Location
Chr.14: 61177614 - 61184010 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977538.1 XP_027833339.1 3 - 4 461 57

View Full Product Details