Assay Design



Gene Transcripts

Gene Symbol
ALDH1A1
Entrez Gene ID
443343
Gene Name
aldehyde dehydrogenase 1 family member A1
Gene Aliases
-
Chromosome Location
Chr.2: 68108099 - 68163247 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009778.1 NP_001009778.1 7 - 8 791 59
GenBank mRNA U12761.1 - 7 - 8 791 59

View Full Product Details