Assay Design



Gene Transcripts

Gene Symbol
ADI1
Entrez Gene ID
101113992
Gene Name
acireductone dioxygenase 1
Gene Aliases
-
Chromosome Location
Chr.2: 1939071 - 1947363 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964044.1 XP_027819845.1 3 - 4 471 55

View Full Product Details