Assay Design



Gene Transcripts

Gene Symbol
IL13
Entrez Gene ID
664707
Gene Name
interleukin 13
Gene Aliases
-
Chromosome Location
Chr.5: 20223835 - 20226665 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082594.1 NP_001076063.1 2 - 3 238 76
GenBank mRNA DQ679798.1 - 2 - 3 238 76

View Full Product Details