Assay Design



Gene Transcripts

Gene Symbol
MYF5
Entrez Gene ID
443159
Gene Name
myogenic factor 5
Gene Aliases
-
Chromosome Location
Chr.3: 124507558 - 124510904 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015094556.2 XP_014950042.1 1 - 2 785 77
GenBank mRNA AF434668.1 - 1 - 2 405 77

View Full Product Details