Assay Design



Gene Transcripts

Gene Symbol
SNCA
Entrez Gene ID
100294604
Gene Name
synuclein alpha
Gene Aliases
-
Chromosome Location
Chr.6: 39617768 - 39779489 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252184.1 NP_001239113.1 2 - 3 164 92
XM_012179343.2 XP_012034733.1 3 - 4 731 92
XM_012179344.3 XP_012034734.1 3 - 4 357 92
XM_012179345.2 XP_012034735.1 3 - 4 350 92
XM_012179346.3 XP_012034736.1 3 - 4 332 92
XM_012179347.3 XP_012034737.1 3 - 4 344 92
XM_012179348.2 XP_012034738.1 3 - 4 346 92
XM_012179350.3 XP_012034740.1 3 - 4 214 92
XM_015096244.2 XP_014951730.1 3 - 4 342 92
GenBank mRNA JN088689.1 - 2 - 3 164 92

View Full Product Details