Assay Design



Gene Transcripts

Gene Symbol
POLE3
Entrez Gene ID
101106138
Gene Name
DNA polymerase epsilon 3, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.2: 10638685 - 10641601 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003991.3 XP_004004040.1 1 - 2 410 110

View Full Product Details