Assay Design



Gene Transcripts

Gene Symbol
TMEM97
Entrez Gene ID
101103331
Gene Name
transmembrane protein 97
Gene Aliases
-
Chromosome Location
Chr.11: 43562188 - 43570563 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012491.4 XP_004012540.2 2 - 3 353 79

View Full Product Details