Assay Design



Gene Transcripts

Gene Symbol
BPGM
Entrez Gene ID
101121089
Gene Name
bisphosphoglycerate mutase
Gene Aliases
-
Chromosome Location
Chr.4: 107102995 - 107134557 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008078.4 XP_004008127.1 2 - 3 780 72
XM_012177164.3 XP_012032554.1 3 - 4 1002 72

View Full Product Details