Assay Design



Gene Transcripts

Gene Symbol
CLEC3B
Entrez Gene ID
101120024
Gene Name
C-type lectin domain family 3 member B
Gene Aliases
-
Chromosome Location
Chr.19: 56089966 - 56096170 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018522.3 XP_004018571.2 2 - 3 426 64

View Full Product Details